missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10711-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Description
The protein encoded by this gene is a DNA binding and apoptosis-inducing protein and is localized in the nucleus. It is also a Rac3-interacting protein which acts as a corepressor for the thyroid hormone receptor. This protein is thought to regulate TRAX/Translin complex formation. Alternate splicing results in multiple transcript variants that encode the same protein. Multiple pseudogenes of this gene are found on chromosome 10.Specifications
| C1D | |
| Polyclonal | |
| Unconjugated | |
| C1D | |
| C1D, hC1D, LRP1, SUN CoR, SUNCOR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10438, 289810, 57316 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O35473, Q13901 | |
| C1d | |
| C1D Fusion Protein Ag1121 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction