Learn More
C15orf27 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26094-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
| C15orf27 | |
| Polyclonal | |
| Unconjugated | |
| TMEM266 | |
| C15orf27, Transmembrane protein C15orf27 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 123591, 244886 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q2M3C6, Q8BZB3 | |
| Tmem266 | |
| C15orf27 Fusion Protein Ag23390 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |