missing translation for 'onlineSavingsMsg'
Learn More

C10orf90 Antibody, Novus Biologicals™

Código de producto. p-200074200 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
25 μL
Tamaño de la unidad:
0.10 ml
25 microlitros
Código de producto. Cantidad unitSize
18636395 25 μL 25 microlitros
18186637 0.1 mL 0.10 ml
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Product Code. 18636395

Brand: Novus Biologicals NBP23883025ul

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

C10orf90 Polyclonal specifically detects C10orf90 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno C10orf90
Aplicaciones Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulación PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
N.º de referencia del gen Q96M02
Alias de gen bA422P15.2, chromosome 10 open reading frame 90, FATS, fragile-site associated tumor suppressor, RP11-422P15.2
Símbolos de los genes C10ORF90
Especie del huésped Rabbit
Inmunógeno This antibody was developed against a recombinant protein corresponding to amino acids: PGLQESHLKETPSLRRVHLGTGACPWSGSFPLENTELANVGANQVTVRKGEKDHTTHCHASDHANQLSIHIPGWSYRAVHTKVFSGSSKRQQGEVCM
Método de purificación Affinity Purified
Cantidad 25 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 118611
Especificidad de la prueba Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.