Descripción
c-Myc Polyclonal antibody specifically detects c-Myc in Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry.
Especificaciones
Especificaciones
| Aplicaciones | Western Blot, ELISA, Immunofluorescence |
| Especie del huésped | Goat |
| Inmunógeno | Goats were immunized with a synthetic peptide representing amino acid residues 410-419 (EQKLISEEDL) of human c-Myc Antibody conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support. |
| Cantidad | 0.1 mL |
| Estado normativo | RUO |
For Research Use Only