Learn More
c-Myc Antibody - BSA Free, Novus Biologicals™
Código de producto. 18745723 Tienda Bio Techne Productos
Código de producto. 18745723 Proveedor Novus Biologicals N.º de proveedor NB600334
Chicken Polyclonal Antibody
c-Myc Polyclonal specifically detects c-Myc in Mouse, Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
For Research Use Only