missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
c-jun Monoclonal antibody specifically detects c-jun in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
Specifications
| Antígeno | c-jun |
| Aplicaciones | Western Blot, Immunoprecipitation |
| Clasificación | Monoclonal |
| Clon | SR1729 |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:1000-1:2000, Immunoprecipitation 1:20 |
| Formulación | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Alias de gen | Activator protein 1, AP1, AP-1, c-Jun, enhancer-binding protein AP1, Jun activation domain binding protein, jun oncogene, jun proto-oncogene, Proto-oncogene c-Jun, transcription factor AP-1, V-jun avian sarcoma virus 17 oncogene homologp39, v-jun sarcoma virus 17 oncogene homolog, v-jun sarcoma virus 17 oncogene homolog (avian) |
| Especie del huésped | Rabbit |
| Inmunógeno | A synthesized peptide derived from human phospho-c-Jun [Thr91] (Uniprot #: P05412) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?