missing translation for 'onlineSavingsMsg'
Learn More

BTN3A1/2/3 Antibody (849203), CoraFluor™ 1, Novus Biologicals™

Product Code. 30154959 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30154959 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30154959 Supplier Novus Biologicals Supplier No. FAB7136CL1

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BTN3A1/2/3 Monoclonal antibody specifically detects BTN3A1/2/3 in Human samples. It is validated for Flow Cytometry,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antígeno BTN3A1/2/3
Aplicaciones Flow Cytometry, CyTOF
Clasificación Monoclonal
Clon 849203
Conjugado CoraFluor 1
Formulación PBS
Alias de gen BT3.1;BTF5;BTN3.1;BTN3A1;CD277
Especie del huésped Mouse
Inmunógeno Chinese hamster ovary cell line CHO-derived recombinant human CD277/BTN3A1, Met1-Gly254, Accession # O00481
Método de purificación Protein A or G purified
Cantidad 0.1 mL
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark. Do not freeze.
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.