Learn More
Novus Biologicals™ BST2 Antibody (SBI Biotech patent anti-BST2) - Humanized, Novus Biologicals™
Código de producto. 30046721 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
1 mg
100 μg
50 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
50 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
30046721 1 mg 1 miligramo
30045941 50 μg 50 microgramos
30044154 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 30046721 Proveedor Novus Biologicals™ N.º de proveedor NBP3280071mg
Human Monoclonal Antibody
BST2 Monoclonal antibody specifically detects BST2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno BST2
Aplicaciones Flow Cytometry, ELISA, Functional Assay
Clasificación Monoclonal
Clon SBI Biotech patent anti-BST2
Conjugado Unconjugated
Formulación Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
N.º de referencia del gen Q10589
Alias de gen bone marrow stromal antigen 2, bone marrow stromal cell antigen 2, BST-2, CD317, CD317 antigen, HM1.24 antigen, NPC-A-7, PDCA-1, TETHERIN
Especie del huésped Human
Inmunógeno BST2/ CD317
Método de purificación Protein A purified
Cantidad 1 mg
Estado normativo RUO
Disciplina de investigación Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 684
Reconstitución Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Especies diana Human
Contenido y almacenamiento Store at -20°C in powder form. Store at -80°C once reconstituted.
Formulario Purified
Isotype IgG1