missing translation for 'onlineSavingsMsg'
Learn More

BRCC3 Antibody, Novus Biologicals™

Código de producto. 18254145 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μL
25 μL
Tamaño de la unidad:
100 microlitros
25 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18254145 100 μL 100 microlitros
18655816 25 μL 25 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18254145 Proveedor Novus Biologicals N.º de proveedor NBP258298

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

BRCC3 Polyclonal specifically detects BRCC3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno BRCC3
Aplicaciones Immunocytochemistry, Immunofluorescence
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Formulación PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Alias de gen BRCA1/BRCA2-containing complex subunit 3, BRCA1/BRCA2-containing complex subunit 36, BRCA1/BRCA2-containing complex, subunit 3, BRCA1-A complex subunit BRCC36, BRCC36C6.1ACXorf53, BRISC complex subunit BRCC36, c6.1A, chromosome X open reading frame 53, EC 3.1.2.15, EC 3.4.19.-, lys-63-specific deubiquitinase BRCC36
Símbolos de los genes BRCC3
Especie del huésped Rabbit
Inmunógeno This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PSHVDVRTQAMYQMMDQGFVGLIFSCFIEDKNTKTGRVLYTCFQSIQAQKSSEYERIEIPIHIVPHVTIGKVCLESAVELPKILCQEEQDAYRRIHSLTHLDSVTKIHNGSVFTKNLCSQMSAVSGPLLQWLEDRLE
Método de purificación Affinity Purified
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 79184
Especificidad de la prueba Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Especies diana Human
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Mostrar más Mostrar menos

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.