missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Bovine FGF acidic/FGF1 Protein
Descripción
The Bovine FGF acidic Protein is derived from Bovine Brain. The Bovine FGF acidic Protein has been validated for the following applications: Bioactivity.
Especificaciones
Especificaciones
| Formulación | Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl with BSA as a carrier protein. |
| ID de gen (Entrez) | 281160 |
| Nombre | FGF acidic |
| Cantidad | 25 μg |
| Fuente | Bovine brain tissue-derived FGF acidic/FGF1 protein |
| Requisitos de almacenamiento | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Alias de gen | AFGF, alpha, alpha-ECGF, beta-ECGF, ECGF, ECGFB, ECGF-betaAcidic fibroblast growth factor, endothelial cell growth factor, beta, FGF acidic, FGF-1, FGFABeta-endothelial cell growth factor, FGF-alpha, fibroblast growth factor 1 (acidic), GLIO703, HBGF1, HBGF-1, heparin-binding growth factor 1 |
| Conjugado | Unconjugated |
| Grado de pureza o calidad | >97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?