Learn More
BORIS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 11074-2-AP-150UL
 Ver más versiones de este producto
Código de producto. 16877903
  • 410.00€ / 150 microlitros
Descripción
Polyclonal anti-CFCFL antibody (11074-2-AP) is produced by immunizing animals with part of C-terminal chain of CTCFL and detects a 75 kDa and a 55 kDa band.
CTCFL is a paralog of CTCF and appears to be expressed primarily in the cytoplasm of spermatocytes. CTCFL is normally expressed in a mutually exclusive pattern that correlates with resetting of methylation marks during male germ cell differentiation.
Especificaciones
Especificaciones
BORIS
Polyclonal
Unconjugated
CTCFL
BORIS, Cancer/testis antigen 27, CCCTC binding factor, CT27, CTCF like protein, CTCF paralog, CTCF T, CTCFL, Zinc finger protein CTCF T
Rabbit
Antigen Affinity Chromatography
RUO
100360757, 140690, 664799
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A2APF3, Q8NI51
Ctcfl
BORIS Fusion Protein Ag1547
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones