Learn More
Invitrogen™ Biotin-Ubiquitin
Código de producto. p-7075648
Click to view available options
Cantidad:
100 μg
10 μg
Tamaño de la unidad:
10 microgramos
100 microgramos
Código de producto. 11545016
Marca: Invitrogen™ PV4379
Biotin-labeled ubiquitin for use as a general research tool, or in high throughput assays to monitor protein ubiquitination.
Biotin-labeled ubiquitin for use as a general research tool, or in high throughput assays to monitor protein ubiquitination. Due to the method in labeling the ubiquitin protein, all of the lysines with the protein are available for further conjugation to form poly-ubiquitin chains.
For use in combination with other LanthaScreen™ products including LanthaScreen™ Tb-Ubiquitin (Cat. No. PV4375), Fluorescein-Ubiquitin (PV4377), or a LanthaScreen™ antibody such as Tb-anti-biotin (PV4749).
TRUSTED_SUSTAINABILITY
Especificaciones
Cantidad 10 μg
Conjugado Biotin
Contenido y almacenamiento Biotin labeled ubiquitin in HEPES buffered saline. Store at -20°C.
Método de detección Fluorescence
Excitación/emisión 340/490, 520 nm
Para utilizar con (equipo) Microplate Reader
Etiqueta o tinte Biotin
Tipo de etiqueta Biotin & Other Haptens
Presentación de datos End Point
Condiciones de envío Dry Ice
Propiedades de sustrato Protein-Based Substrate
Técnica TR-FRET
Formulario Liquid
Mostrar más Mostrar menos
For Research Use Only. Not for use in diagnostic procedures.