missing translation for 'onlineSavingsMsg'
Learn More

BIN2 Antibody, Novus Biologicals™

Product Code. p-7109472 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
100 μL
Unit Size:
100 microlitros
Product Code. Cantidad unitSize
18264777 100 μL 100 microlitros
1 options
This item is not returnable. View return policy

Product Code. 18264777

Brand: Novus Biologicals NBP179648

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BIN2 Polyclonal specifically detects BIN2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno BIN2
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.5 mg/ml
Conjugado Unconjugated
Dilución Western Blot 1.0 ug/ml
Formulación PBS, 2% Sucrose with 0.09% Sodium Azide
N.º de referencia del gen NP_057377
Alias de gen BRAP1, BRAP-1, breast cancer associated protein BRAP1, Breast cancer-associated protein 1, bridging integrator 2, bridging integrator-2
Símbolos de los genes BIN2
Especie del huésped Rabbit
Inmunógeno Synthetic peptide directed towards the middle region of human BIN2The immunogen for this antibody is BIN2. Peptide sequence EEVLPSSTTPSPGGALSPSGQPSSSATEVVLRTRTASEGSEQPKKRASIQ.
Peso molecular del antígeno 62 kDa
Método de purificación Affinity purified
Cantidad 100 μL
Estado normativo RUO
Disciplina de investigación Cell Cycle and Replication
Primario o secundario Primary
ID de gen (Entrez) 51411
Especificidad de la prueba Expected identity based on immunogen sequence: Pig: 86%; Bovine: 79%; Rabbit: 79%; Rat: 77%.
Reconstitución Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Especies diana Human, Rat, Pig, Bovine, Canine, Equine, Rabbit
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.