missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ beta Synuclein Monoclonal Antibody (6A10)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA547455
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
beta Synuclein Monoclonal Antibody for Western Blot, ICC/IF, IHC (PFA)
Synuclein family of proteins is abundantly expressed in neuronal cytosol and presynaptic terminals. In vertebrates they are encoded by three different genes. Synucleins have been specifically implemented in three major diseases: Alzheimer's (AD), Parkinson's PD) and breast cancer. In AD, a peptide derived from alpha synuclein forms an intrinsic component of plaque amyloid. In PD, an alpha synuclein accumulates in Lewy bodies. An allele of alpha synuclein has been linked to many familial cases of PD. In breast cancer increased expression of gamma synuclein correlates with the disease progression. Synucleins appear to be involved in the membrane plasticity in developing song control system of songbirds.
Especificaciones
| beta Synuclein | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 5mM sodium azide | |
| P33567, Q16143, Q63754, Q91ZZ3 | |
| SNCB | |
| C-terminal peptide of human beta-synuclein EPEGESYEDPPQEEYQEYEPEA coupled to KLH. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Bovine, Pig | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
| 6A10 | |
| Unconjugated | |
| SNCB | |
| AI838531; betaSYN; beta-synuclein; Phosphoneuroprotein 14; PNP 14; Sncb; synuclein beta; synuclein, beta | |
| Mouse | |
| Protein G | |
| RUO | |
| 100127169, 104069, 113893, 508421, 6620 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?