Learn More
beta-III Tubulin Antibody, Novus Biologicals™
Código de producto. 18715083 Tienda Bio Techne Productos
Código de producto. 18715083
Marca: Novus Biologicals NB1001612
Chicken Polyclonal Antibody has been used in 16 publications
beta-III Tubulin Polyclonal specifically detects beta-III Tubulin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry Whole-Mount, Knockdown Validated.
TRUSTED_SUSTAINABILITY
For Research Use Only