missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
beta COP Polyclonal specifically detects beta COP in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antígeno | beta COP |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/ml |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1.0 ug/ml |
| Formulación | PBS, 2% Sucrose with 0.09% Sodium Azide |
| N.º de referencia del gen | P53618 |
| Alias de gen | beta coat protein, Beta-coat protein, beta-cop, coatomer protein complex, subunit beta, coatomer protein complex, subunit beta 1, coatomer subunit beta, COPBFLJ57957, DKFZp761K102, FLJ10341, FLJ46444 |
| Símbolos de los genes | COPB1 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?