Learn More
Bestrophin 1 Antibody (E6-6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 42 publications
Marca: Novus Biologicals NB300-164
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Bestrophin 1 Monoclonal specifically detects Bestrophin 1 in Human, Mouse, Porcine, Canine, Primate, Rat (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Proximity Ligation Assay, Dual RNAscope ISH-IHC.
Especificaciones
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), In situ PLA | |
| Pig, Primate, Rat (Negative) | |
| Ascites |
| Monoclonal | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido