missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BEGAIN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14956-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
May sustain the structure of the postsynaptic density (PSD).Especificaciones
BEGAIN | |
Polyclonal | |
Unconjugated | |
BEGAIN | |
BEGAIN, KIAA1446 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
380785, 57596, 79146 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O88881, Q68EF6, Q9BUH8 | |
Begain | |
BEGAIN Fusion Protein Ag6794 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |