missing translation for 'onlineSavingsMsg'
Learn More

BCMA/TNFRSF17 Antibody (CD269/8276R), Novus Biologicals™

Product Code. p-200004398 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
100 μg
20 μg
Unit Size:
100 microgramos
20 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
18683465 20 μg 20 microgramos
18652764 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18683465 Supplier Novus Biologicals Supplier No. NBP32043420ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

BCMA/TNFRSF17 Monoclonal antibody specifically detects BCMA/TNFRSF17 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno BCMA/TNFRSF17
Aplicaciones ELISA, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon CD269/8276R
Concentración 0.2 mg/ml
Conjugado Unconjugated
Dilución ELISA, Immunohistochemistry-Paraffin 1-2 μg/mL
Formulación 10mM PBS with 0.05% BSA
Alias de gen B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, tumor necrosis factor receptor superfamily, member 17
Especie del huésped Rabbit
Inmunógeno Recombinant human BCMA/TNFRSF17 protein fragment (around aa 1-184) (exact sequence is proprietary)
Método de purificación Protein A or G purified
Cantidad 20 μg
Estado normativo RUO
Disciplina de investigación Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 608
Especies diana Human
Contenido y almacenamiento Store at 4&drg;C. Do not freeze.
Formulario Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.