Learn More
BCL11B Antibody (CL6426) [HRP], Novus Biologicals™
Código de producto. 30296750 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
30296750 0.1 mL 0.10 ml
1 options
Código de producto. 30296750 Proveedor Novus Biologicals N.º de proveedor NBP276493H
Mouse Monoclonal Antibody
BCL11B Monoclonal antibody specifically detects BCL11B in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno BCL11B
Aplicaciones Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon CL6426
Conjugado HRP
Formulación PBS
Alias de gen B-cell CLL/lymphoma 11B, B-cell CLL/lymphoma 11B (zinc finger protein), B-cell CLL/lymphoma 11B/T-cell receptor delta constant region fusion protein, B-cell lymphoma/leukaemia 11B, B-cell lymphoma/leukemia 11B, BCL-11B, BCL11B/TRDC fusion, COUP-TF-interacting protein 2, CTIP-2, CTIP2ZNF856B, hRit1, hRIT1-alpha, Radiation-induced tumor suppressor gene 1 protein, RIT1, zinc finger protein hRit1 alpha
Especie del huésped Mouse
Inmunógeno This antibody was developed using a synthetic peptide derived from Q9C0K0, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Apoptosis, Cancer, Transcription Factors and Regulators, Tumor Suppressors
Primario o secundario Primary
ID de gen (Entrez) 64919
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG1