Learn More
BAP1 Antibody (BAP1/9795R), Novus Biologicals™
Código de producto. 30288137 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
20 μg
Tamaño de la unidad:
100 microgramos
20 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
30288137 20 μg 20 microgramos
30293741 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 30288137 Proveedor Novus Biologicals N.º de proveedor NBP34462420ug
Rabbit Monoclonal Antibody
BAP1 Monoclonal antibody specifically detects BAP1 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno BAP1
Aplicaciones Immunohistochemistry
Clasificación Monoclonal
Clon BAP1/9795R
Concentración 0.2 mg/ml
Conjugado Unconjugated
Formulación 10 mM PBS with 0.05% BSA
Alias de gen BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase), BRCA1-associated protein 1, Cerebral protein 6, cerebral protein-13, cerebral protein-6, DKFZp686N04275, EC 3.4.19.12, FLJ35406, FLJ37180, hucep-6, KIAA0272HUCEP-13, ubiquitin carboxyl-terminal hydrolase BAP1, ubiquitin carboxy-terminal hydrolase, UCHL2
Especie del huésped Rabbit
Inmunógeno Recombinant fragment (around aa400-729) of human BAP1 protein (Uniprot: Q92560). Exact sequence is proprietary.
Método de purificación Protein A or G purified
Cantidad 20 μg
Estado normativo RUO
Disciplina de investigación Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Tumor Suppressors
Primario o secundario Primary
ID de gen (Entrez) 8314
Especies diana Human
Contenido y almacenamiento Store at 4⪚C.
Formulario Purified
Isotype IgG κ