missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ BAIAP2 Recombinant Protein
Especificaciones
Reactividad cruzada | Human |
---|---|
Formulación | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Peso molecular | 82.06 |
Nombre | BAIAP2 (Human) Recombinant Protein (P01) |
Intervalo de pH | 8 |
Descripción
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Especificaciones
Human | |
82.06 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Yes | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
BAIAP2 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) |