missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BACH2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-58464-25ul
Este artículo no se puede devolver.
Vea la política de devoluciones
Description
BACH2 Polyclonal specifically detects BACH2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| BACH2 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
| BTB and CNC homolog 2, BTB and CNC homology 1, basic leucine zipper transcription factor 2, transcription regulator protein BACH2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 60468 | |
| Human | |
| IgG |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
| BACH2 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SNSLKPGLARGQIKSEPPSEENEEESITLCLSGDEPDAKDRAGDVEMDRKQPSPAPTPTAPAGAACLERSRSVASPSCLRSLFSITK | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction