missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
B7-H6 Monoclonal antibody specifically detects B7-H6 in Cynomolgus Monkey samples. It is validated for Flow Cytometry, CyTOF
Especificaciones
Especificaciones
| Antígeno | B7-H6 |
| Aplicaciones | Flow Cytometry, CyTOF |
| Clasificación | Monoclonal |
| Clon | 1002932 |
| Conjugado | APC |
| Dilución | Flow Cytometry, CyTOF-ready |
| Formulación | PBS |
| Alias de gen | B7 homolog 6, B7-H6, DKFZp686O24166 |
| Especie del huésped | Mouse |
| Inmunógeno | Human embryonic kidney cell, HEK293-derived cynomolgus Monkey B7-H6, Asp25-Ser262 |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?