missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
B7-H2/ICOSLG Polyclonal antibody specifically detects B7-H2/ICOSLG in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Especificaciones
Especificaciones
| Antígeno | B7-H2/ICOSLG |
| Aplicaciones | Immunofluorescence |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL |
| Formulación | PBS (pH 7.2) and 40% Glycerol |
| Alias de gen | B7H2B7 homolog 2, B7-H2B7-related protein 1, B7RP1B7-like protein Gl50, B7RP-1LICOS, CD275, CD275 antigen, GL50transmembrane protein B7-H2 ICOS ligand, ICOSL, ICOS-L, inducible T-cell co-stimulator ligand, KIAA0653ICOS ligand |
| Especie del huésped | Rabbit |
| Inmunógeno | This antibody was developed against a recombinant protein corresponding to amino acids: KEVRAMVGSDVELSCACPEGSRFDLNDVYVYWQTSESKTVVTYHIPQNSSLENVDSRYRNRALMSPAGMLRGDFSLRLFNVT |
| Método de purificación | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?