missing translation for 'onlineSavingsMsg'
Learn More

ATRIP Antibody (OTI1G6), Alexa Fluor™ 405, Novus Biologicals™

Código de producto. 18188724 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18188724 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. View return policy
Product Code. 18188724 Supplier Novus Biologicals Supplier No. NBP272270AF405

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ATRIP Monoclonal specifically detects ATRIP in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno ATRIP
Aplicaciones Western Blot
Clasificación Monoclonal
Clon OTI1G6
Conjugado Alexa Fluor 405
Dilución Western Blot
Formulación 50mM Sodium Borate with 0.05% Sodium Azide
Alias de gen AGS1, ATM and Rad3-related-interacting protein, ATR interacting protein, ATR-interacting protein, DKFZp762J2115, FLJ12343, MGC20625, MGC21482, MGC26740
Símbolos de los genes ATRIP
Especie del huésped Mouse
Inmunógeno Recombinant protein fragment corresponding to amino acids 42-309 of human ATRIP (NP_569055) produced in E.coli.
Método de purificación Affinity Purified
Cantidad 0.1 mL
Disciplina de investigación Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair
Primario o secundario Primary
ID de gen (Entrez) 84126.0
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.