missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
ATP13A2 Polyclonal specifically detects ATP13A2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Especificaciones
Especificaciones
| Antígeno | ATP13A2 |
| Aplicaciones | Western Blot, Immunohistochemistry |
| Clasificación | Polyclonal |
| Conjugado | Alexa Fluor 647 |
| Dilución | Western Blot, Immunohistochemistry |
| Formulación | 50mM Sodium Borate with 0.05% Sodium Azide |
| N.º de referencia del gen | Q9NQ11 |
| Alias de gen | ATPase type 13A2, EC 3.6.3, EC 3.6.3.-, EC 3.6.3.5, EC 3.6.3.8, FLJ26510, HSA9947, KRPPD, PARK9putative ATPase, Parkinson disease (autosomal recessive) 9 (Kufor-Rakeb syndrome), probable cation-transporting ATPase 13A2 |
| Símbolos de los genes | ATP13A2 |
| Especie del huésped | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?