Learn More
ATG16L1 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 29445-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The ATG16L1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ATG16L1. This antibody recognizes human, mouse, rat antigen. The ATG16L1 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Especificaciones
| ATG16L1 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 55054 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| 1000 μg/mL | |
| APG16L; ATG16L; FLJ00045; FLJ10035; FLJ10828; FLJ22677; IBD10; WDR30 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 68-75 kDa | |
| Liquid |