Descripción
Atad5 Polyclonal antibody specifically detects Atad5 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Especificaciones
Especificaciones
| Antígeno | Atad5 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500-1:1000, ELISA |
| Formulación | PBS, 50% Glycerol, pH (7.3) |
| Alias de gen | ATPase family AAA domain-containing protein 5, ATPase family, AAA domain containing 5, C17orf41, chromosome fragility associated gene 1, Chromosome fragility-associated gene 1 protein, ELG1, enhanced level of genomic instability 1 homolog, FLJ12735, FRAG1chromosome 17 open reading frame 41 |
| Especie del huésped | Rabbit |
| Inmunógeno | FGF receptor activating protein 1. (Uniprot# Q96QE3) |
| Método de purificación | Immunogen affinity purified |
| Mostrar más |