Descripción
ASRGL1 Monoclonal antibody specifically detects ASRGL1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | ASRGL1 |
| Aplicaciones | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | CL1679 |
| Conjugado | PE |
| Formulación | PBS |
| Alias de gen | ALP1, ALPL-asparagine amidohydrolase, asparaginase like 1, asparaginase-like 1 protein, Asparaginase-like protein 1, CRASH, EC 3.5.1.1, FLJ22316, L-asparaginase |
| Especie del huésped | Mouse |
| Inmunógeno | This antibody was developed using a recombinant protein derived from Q7L266, with the exact immunogen sequence remaining proprietary. |
| Método de purificación | Protein A purified |
| Mostrar más |