missing translation for 'onlineSavingsMsg'
Learn More

Asialoglycoprotein Receptor 2 Antibody (2327C) [Janelia Fluor™ 669], Novus Biologicals™

Código de producto. 30173617 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30173617 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30173617 Supplier Novus Biologicals Supplier No. FAB9970JF669

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Asialoglycoprotein Receptor 2 Monoclonal antibody specifically detects Asialoglycoprotein Receptor 2 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Asialoglycoprotein Receptor 2
Aplicaciones Flow Cytometry, Immunohistochemistry, Western Blot
Clasificación Monoclonal
Clon 2327C
Conjugado Janelia Fluor 669
Formulación 50mM Sodium Borate
Alias de gen ASGPR 2, ASGP-R 2, ASGPR2, ASGP-R2, asialoglycoprotein receptor 2, CLEC4H2FLJ60040, C-type lectin domain family 4 member H2, HBxAg-binding protein, HBXBP, Hepatic lectin H2, HL-2
Especie del huésped Rabbit
Inmunógeno Chinese hamster ovary cell line CHO-derived recombinant human ASGR2, Gln80-Ala311, Accession # P07307
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 433
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.