missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beskrivelse
ASAP1 Polyclonal antibody specifically detects ASAP1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Tekniske data
Tekniske data
| Antígeno | ASAP1 |
| Aplicaciones | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Formulación | PBS (pH 7.2), 40% Glycerol |
| Alias de gen | 130 kDa phosphatidylinositol 4,5-biphosphate-dependent ARF1 GTPase-activatingprotein, AMAP1, ARF GTPase-activating protein 1, arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 1, ArfGAP with SH3 domain, ankyrin repeat and PH domain 1, centaurin, beta 4, CENTB4, DDEF1ADP-ribosylation factor-directed GTPase-activating protein 1, development and differentiation enhancing factor 1, Development and differentiation-enhancing factor 1, Differentiation-enhancing factor 1, KIAA1249DEF-1, PAG2, PAP, PIP2-dependent ARF1 GAP, ZG14P |
| Especie del huésped | Rabbit |
| Inmunógeno | This antibody was developed against a recombinant protein corresponding to amino acids: STPRDKQRLSYGAFTNQIFVSTSTDSPTSPTTEAPPLPPRNAGKGNDGGPSSSSKTTNKFEGLSQQSSTSSAKTALGPRVLPKLPQKVALRKT |
| Método de purificación | Immunogen affinity purified |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?