missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ART1 Polyclonal antibody specifically detects ART1 in Mouse samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antígeno | ART1 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Immunohistochemistry-Paraffin 1:500-1:2000 |
| Formulación | 0.2 μm filtered solution in PBS |
| Alias de gen | ADP-ribosyltransferase 1, ADP-ribosyltransferase 2, CD296, GPI-linked NAD(P)(+)--arginine ADP-ribosyltransferase 1, MGC133217, mono(ADP-ribosyl)transferase 1, RT6 |
| Especie del huésped | Rabbit |
| Inmunógeno | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the mouse ART1. |
| Método de purificación | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?