missing translation for 'onlineSavingsMsg'
Learn More

ARHGAP25 Antibody (OTI2D5), DyLight 350, Novus Biologicals™

Código de producto. 18179972 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18179972 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. View return policy
Product Code. 18179972 Supplier Novus Biologicals Supplier No. NBP271508UV

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ARHGAP25 Monoclonal specifically detects ARHGAP25 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno ARHGAP25
Aplicaciones Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Clasificación Monoclonal
Clon OTI2D5
Conjugado DyLight 350
Dilución Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulación 50mM Sodium Borate with 0.05% Sodium Azide
Alias de gen KAIA0053, KIAA0053, Rho GTPase activating protein 25, rho GTPase-activating protein 25, Rho-type GTPase-activating protein 25
Símbolos de los genes ARHGAP25
Especie del huésped Mouse
Inmunógeno Full length recombinant protein of human ARHGAP25 (NP_055697) produced in HEK293T cells.
Método de purificación Affinity Purified
Cantidad 0.1 mL
Primario o secundario Primary
ID de gen (Entrez) 9938.0
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.