missing translation for 'onlineSavingsMsg'
Learn More

Arginase 1/ARG1/liver Arginase Antibody (OTI4E6), Janelia Fluor™ 646, Novus Biologicals™

Código de producto. 18133704 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18133704 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18133704 Proveedor Novus Biologicals N.º de proveedor NBP270204JF646

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Arginase 1/ARG1/liver Arginase Monoclonal specifically detects Arginase 1/ARG1/liver Arginase in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Arginase 1/ARG1/liver Arginase
Aplicaciones Western Blot, Flow Cytometry, Immunohistochemistry
Clasificación Monoclonal
Clon OTI4E6
Conjugado Janelia Fluor 646
Dilución Western Blot, Flow Cytometry, Immunohistochemistry
Formulación 50mM Sodium Borate with 0.05% Sodium Azide
Alias de gen arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, Liver-type arginase, Type I arginase
Símbolos de los genes ARG1
Especie del huésped Mouse
Inmunógeno Full length recombinant protein of human Arginase 1/ARG1/liver Arginase (NP_000036) produced in HEK293T cells.
Método de purificación Affinity Purified
Cantidad 0.1 mL
Disciplina de investigación Cancer, Cellular Markers, Chromatin Research, Lipid and Metabolism, Myeloid derived Suppressor Cell
Primario o secundario Primary
ID de gen (Entrez) 383.0
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.