missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Aquaporin 4 Monoclonal Antibody (C9), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen™ MA543954
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
In skeletal muscle, AQP4 (aquaporin 4 also known as mercurial insensitive water channel), localizes to the sarcolemma of fast-twitch muscle fibers. Aquaporins (AQPs) are a large family of integral membrane water transport channel proteins that facilitate the transport of water through the cell membrane. This function is conserved in animals, plants and bacteria. Many isoforms of aquaporin have been identified in mammals, designated AQP0 through AQP10. Aquaporins are widely distributed and it is not uncommon for more than one type of AQP to be present in the same cell. Although most aquaporins are only permeable to water, AQP3, AQP7, AQP9 and one of the two AQP10 transcripts are also permeable to urea and glycerol.
Especificaciones
| Aquaporin 4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P47863, P55087 | |
| AQP4 | |
| Cys178-Gly317 (Accession P55087), with N-terminal His and GST Tag. | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2a κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| C9 | |
| Unconjugated | |
| AQP4 | |
| Aqp4; AQP-4; aquaporin 4; aquaporin type4; Aquaporin4; aquaporin-4; HMIWC2; Mercurial-insensitive water channel; MGC22454; MIWC; MIWC2; WCH4 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 25293, 361 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido