Learn More
APP Antibody (S05-3E3), Novus Biologicals™
Código de producto. 18635574 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
50 μg
Tamaño de la unidad:
100 microlitros
50 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18635574 50 μg 50 microlitros
18644115 100 μg 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18635574 Proveedor Novus Biologicals N.º de proveedor NBP32001550ul
Rabbit Monoclonal Antibody
APP Monoclonal antibody specifically detects APP in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno APP
Aplicaciones Western Blot
Clasificación Monoclonal
Clon S05-3E3
Concentración 0.3 mg/mL
Conjugado Unconjugated
Formulación 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Alias de gen amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP
Especie del huésped Rabbit
Inmunógeno A synthetic peptide of human APP (Uniprot # P05067)
Método de purificación Affinity purified
Cantidad 50 μg
Estado normativo RUO
Disciplina de investigación Alzheimers Research, Apoptosis, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific
Primario o secundario Primary
ID de gen (Entrez) 351
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG