missing translation for 'onlineSavingsMsg'
Learn More

Apolipoprotein D Antibody (APOD/3412), FITC, Novus Biologicals™

Código de producto. 18680408 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18680408 100 μg 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18680408 Proveedor Novus Biologicals N.º de proveedor NBP308417F

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Apolipoprotein D Monoclonal antibody specifically detects Apolipoprotein D in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Apolipoprotein D
Aplicaciones Immunohistochemistry (Paraffin), Protein Array
Clasificación Monoclonal
Clon APOD/3412
Conjugado FITC
Formulación PBS
Alias de gen apo-D, ApoD, apolipoprotein D
Especie del huésped Mouse
Inmunógeno Human recombinant Apolipoprotein D protein fragment (around aa 21-119) (exact sequence is proprietary) (Uniprot: P05090)
Método de purificación Protein A or G purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación Alzheimers Research, Cancer, Cardiovascular Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 347
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.