Learn More
ApoE4 Antibody (4E4), PE, Novus Biologicals™
Código de producto. 18542804 Tienda Bio Techne Productos
Código de producto. 18542804
Marca: Novus Biologicals NBP149529PE
Mouse Monoclonal Antibody
ApoE4 Monoclonal antibody specifically detects ApoE4 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation