missing translation for 'onlineSavingsMsg'
Learn More

APOB48R Antibody, FITC, Novus Biologicals™

Product Code. 18663052 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
Product Code. Cantidad unitSize
18663052 0.1 mL 0.10 ml
1 options
This item is not returnable. View return policy

Product Code. 18663052

Brand: Novus Biologicals NBP298580F

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

APOB48R Polyclonal antibody specifically detects APOB48R in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno APOB48R
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado FITC
Formulación PBS
Alias de gen APOB100R, apoB-48R, APOB48Rapolipoprotein B48 receptor, apolipoprotein B receptor, Apolipoprotein B-100 receptor, Apolipoprotein B-48 receptor
Especie del huésped Rabbit
Inmunógeno Produced in rabbits immunized with E. coli-derived Human APOB48R fragment.
Método de purificación Antigen and protein A Affinity-purified
Cantidad 0.1 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 55911
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.