missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
AP2 alpha Monoclonal antibody specifically detects AP2 alpha in Human samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Western Blot,Radioimmune Assays,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
Specifications
| Antígeno | AP2 alpha |
| Aplicaciones | Flow Cytometry, ELISA, Immunohistochemistry, Western Blot, Radioimmune Assays (RIA) |
| Clasificación | Monoclonal |
| Clon | A6/2/2 |
| Conjugado | CoraFluor 1 |
| Formulación | PBS |
| Alias de gen | Activating enhancer-binding protein 2-alpha, Activator protein 2, AP-2 transcription factor, AP2-alpha, AP2TFAP-2alpha, AP-2transcription factor AP-2 alpha (activating enhancer-binding protein 2 alpha), BOFS, FLJ51761, TFAP2, transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha), transcription factor AP-2-alpha |
| Especie del huésped | Mouse |
| Inmunógeno | Peptide to C-Terminal of AP2 alpha coupled to KLH. PNSHTDNNAKSSDKEEKHRK |
| Método de purificación | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?