Learn More
WARS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA529102
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: HepG2, mouse brain. Predicted reactivity: Mouse (94%), Rat (93%), Zebrafish (86%), Xenopus laevis (89%), Rabbit (93%), Chimpanzee (100%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Two forms of tryptophanyl-tRNA synthetase exist, a cytoplasmic form, named WARS, and a mitochondrial form, named WARS2. Tryptophanyl-tRNA synthetase (WARS) catalyzes the aminoacylation of tRNA(trp) with tryptophan and is induced by interferon. Tryptophanyl-tRNA synthetase belongs to the class I tRNA synthetase family. Four transcript variants encoding two different isoforms have been found for this gene.Especificaciones
WARS | |
Polyclonal | |
Unconjugated | |
WARS1 | |
GAMMA-2; hWRS; IFI 53; IFI53; IFP 53; IFP53; Interferon-induced protein 53; T1-TrpRS; T2-TrpRS; TrpRS; tryptophan tRNA ligase 1, cytoplasmic; tryptophan--tRNA ligase, cytoplasmic; Tryptophanyl-tRNA synthetase; tryptophanyl-tRNA synthetase, cytoplasmic; Wars; WARS protein; Wrs | |
Rabbit | |
Protein A | |
RUO | |
22375, 314442, 7453 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P23381, P32921, Q6P7B0 | |
WARS1 | |
Recombinant fragment corresponding to a region within amino acids 151 and 430 of Human Tryptophanyl tRNA synthetase | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |