Learn More
SAP130 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA529450
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (98%), Xenopus laevis (83%), Chicken (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SAP130 is a subunit of the histone deacetylase-dependent SIN3A corepressor complex (Fleischer et al., 2003).Especificaciones
SAP130 | |
Polyclonal | |
Unconjugated | |
Sap130 | |
130 kDa Sin3-associated polypeptide; 2610304F09Rik; 6720406D06; FLJ12761; Histone deacetylase complex subunit SAP130; KIAA0017; mSin3A-associated protein 130; pre-mRNA splicing factor SF3b, 130 kDa subunit; pre-mRNA-splicing factor SF3b 130 kDa subunit; RGD1311657; RSE1; SAP 130; SAP130; SF3b130; SF3B3; Sin3A associated protein; Sin3A associated protein 130; Sin3A associated protein 130kDa; Sin3A associated protein, 130kDa; Sin3A-associated protein, 130kDa; Sin3-associated polypeptide p130; spliceosome-associated protein 130; Splicing factor 3B subunit 3; STAF130 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79595 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9H0E3 | |
Sap130 | |
Recombinant fragment corresponding to a region within amino acids 771 and 1045 of Human SAP130 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |