missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPY4R Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA534104
Detalles adicionales : Peso : 0.01000kg
Descripción
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Horse (100%) Bovine, Pig (94%) Bat, Panda, Rabbit (88%) Dog (81%).
Receptor for neuropeptide Y and peptide YY. The rank order of affinity of this receptor for pancreatic polypeptides is PP, PP (2-36) and [Ile-31, Gln-34] PP > [Pro-34] PYY > PYY and [Leu-31, Pro-34] NPY > NPY > PYY (3-36) and NPY (2-36) > PP (13- 36) > PP (31-36) > NPY free acid.Especificaciones
NPY4R | |
Polyclonal | |
Unconjugated | |
NPY4R | |
neuropeptide Y receptor type 4; neuropeptide Y receptor Y4; NPY4R; NPY4-R; NPYR-D; NYYR-D; Pancreatic polypeptide receptor 1; PP1; PPYR1; Y4; Y4 receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5540 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P50391 | |
NPY4R | |
Synthetic 16 amino acid peptide from 2nd extracellular domain of human PPYR1. | |
50 μg | |
Primary | |
Equine, Human, Non-human Primate | |
Antibody | |
IgG |