Learn More
Phospho-IRS1 (Tyr612) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen 44816G
368.00 EUR hasta el 2024-06-28
Use el código promocional "21649" para aplicar la oferta.
Detalles adicionales : Peso : 0.01000kg
Descripción
On western blotting this antibody detects a protein with the apparent MW of 165 kDa. Positive controls used were Insulin (50 nM for 5 minutes) or TPA (100 ng/mL for 30 min) treated human embryonic kidney cells (293T) or Chinese Hamster Ovary cells (CHO-T) transiently transfected with a plasmid encoding human IRS1.
Phospho-IRS1 (Tyr612) Polyclonal antibody specifically detects Phospho-IRS1 (Tyr612) in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Especificaciones
Phospho-IRS1 (Tyr612) | |
Polyclonal | |
Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3 | |
P35568 | |
IRS1 | |
The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human IRS-1 that contains tyrosine 612. The sequence is conserved in mouse, rat and chicken. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
IRS1 | |
G972R; HIRS-1; Insulin receptor substrate 1; IRS1; IRS-1; IRS1IRM; pp185 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3667 | |
-20°C | |
Liquid |