Learn More
IL-17A Rat anti-Mouse, Unlabeled, Clone: TC11-18H10, BD
Rat Monoclonal Antibody
Marca: BD Biosciences 555068
Detalles adicionales : Peso : 0.01000kg
Descripción
The TC11-18H10 antibody reacts with recombinant and natural mouse IL-17A proteins. IL-17A, also known as CTLA-8, is a T cell-derived cytokine that promotes inflammatory responses. Mouse IL-17A is a proinflammatory cytokine that can induce the release of IL-6 by mouse stromal cells. It has been shown to support the growth of hemopoietic progenitors in vitro; it can also stimulate granulopoiesis in vivo. The TC11-18H10 antibody has been reported to neutralize IL-17A activity. Recent studies have shown that IL-17A is produced by a unique subset of Th17 cells that develop along a pathway distinct from the Th1- and Th2- cell differentiation pathways. The mouse IL-17A cDNA was isolated from a cDNA library generated from TCRαβ+CD4-CD8-thymocytes.
Blocking Assay, ELISA, Intracellular Staining, Western Blotting
Especificaciones
IL-17A | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Recombinant Mouse IL-17 Protein | |
0.5 mg | |
Immunology | |
Mouse | |
IgG1 κ |
Flow Cytometry, ELISA, Blocking Assay | |
TC11-18H10 | |
Unconjugated | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
For Research Use Only.