Learn More
Anti-Human CD55 (JS11), Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 65271-1-IG-500UG
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The CD55 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD55. This antibody recognizes human antigen. The CD55 antibody has been validated for the following applications: FC analysis.Especificaciones
| CD55 | |
| Monoclonal | |
| 0.5 mg/mL | |
| CR; CROM; DAF; TC | |
| Affinity purification | |
| RUO | |
| 1604 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry | |
| JS11 | |
| Unconjugated | |
| Mouse | |
| 500 μg | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG1 |