missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXD10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA546861
Detalles adicionales : Peso : 0.00010kg
Descripción
Peptide sequence: NPEVPVPGYF RLSQTYATGK TQEYNNSPEG SSTVMLQLNP RGAAKPQLSA Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 92%
HOXD10 Polyclonal antibody specifically detects HOXD10 in Human samples. It is validated for Western Blot.Especificaciones
HOXD10 | |
Polyclonal | |
Unconjugated | |
HOXD10 | |
AI385591; AI874987; d10; homeo box 4D; homeo box D10; homeo box D10a; homeobox D10; homeobox D10a; homeobox gene D-10; homeobox protein Hox-4.5; Homeobox protein Hox-4D; homeobox protein Hox-4E; Homeobox protein Hox-5.3; homeobox protein Hox-C10; Homeobox protein Hox-D10; homeobox protein Hox-D10a; homeobox protein Hox-D11; homeobox protein Hox-D10; HOX4; Hox-4.4; Hox-4.5; Hox-45; HOX4D; HOX4E; Hox-5.3; hoxc10; hox-C10; Hoxc10 protein; Hoxd10; HOX-D10; hoxd-10; hoxd10a; hoxd10a protein; z-82 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3236 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P28358 | |
HOXD10 | |
Synthetic peptide directed towards the middle region of human HOXD10 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |