missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat IgG Fraction to Human Fibrinogen, FITC, Polyclonal, MP Biomedicals
Marca: MP Biomedicals 0855169
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Host Species: Goat
Species Reactivity: Human
Especificaciones
Fibrinogen | |
FITC | |
2 mL | |
Human |
Polyclonal | |
Goat | |
Primary | |
Purified |