Learn More
CD48 Hamster anti-Mouse, PE, Clone: HM48-1, BD
Hamster Monoclonal Antibody
Marca: BD Biosciences 557485
Detalles adicionales : Peso : 0.01000kg
Descripción
The HM48-1 monoclonal antibody specifically binds to CD48 (previously known as BCM1 in mice, Blast-1 in human, and OX-45 in the rat), a GPI-anchored member of the Ig superfamily. It is widely distributed on leukocytes, but not on non-hematopoietic cells, and its ligands include CD2 (LFA-2) and CD244 (2B4 antigen). The HM48-1 mAb blocks binding of soluble CD2 to CD48-bearing cells, blocks the interaction of CD2 and CD244, inhibits spleen cell proliferative responses to mitogens, augments the proliferative response of spleen cells when cross-linked with anti-CD3e mAbs, and inhibits priming of CTL in vitro. In vivo administration of HM48-1 antibody can prolong survival of cardiac allografts, an effect which is greatly enhanced by the addition of anti-CD2 mAb 12-15. This hamster mAb to a mouse leukocyte antigen does not cross-react with rat leukocytes.
Especificaciones
CD48 | |
Monoclonal | |
0.2mg/mL | |
BLAST; BLAST-1; BCM1; HM48-1; MEM-102; Sgp-60; SLAMF2 | |
Hamster | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
HM48-1 | |
PE | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse T lymphoma MBL-2 | |
0.1 mg | |
Immunology | |
Mouse | |
IgG1 λ3 |
For Research Use Only.